ATC311765306 Atlantic Research Chemicals Ltd

9-Oxo-9H-thioxanthene-3-carboxylic acid 10,10-dioxide

  • MDL number MFCD00075362
  • Cas Number : 51762-52-8
  • Molecular Weight : 288.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA100124
Availability details for
Quantity
  • Availability
  • Add to Cart