ATC311765308 Atlantic Research Chemicals Ltd

(11-Oxo-6,11-dihydrodibenzo[b,e]oxepin-2-yl)acetic acid

  • MDL number MFCD00242952
  • Cas Number : 55453-87-7
  • Molecular Weight : 268.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA100126
Availability details for
Quantity
  • Availability
  • Add to Cart