ATC311765310 Atlantic Research Chemicals Ltd

(7-Methoxy-1-naphthyl)acetic acid

  • MDL number MFCD12756806
  • Cas Number : 6836-22-2
  • Molecular Weight : 216.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA100128
Availability details for
Quantity
  • Availability
  • Add to Cart