ATC311765316 Atlantic Research Chemicals Ltd

3-Amino-6-methyl-4-(trifluoromethyl)thieno[2,3-b]pyridine-2-carboxylic acid

  • MDL number MFCD01055887
  • Cas Number : 610259-30-8
  • Molecular Weight : 276.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA100133
Availability details for
Quantity
  • Availability
  • Add to Cart