ATC311765376 Atlantic Research Chemicals Ltd

Tetrahydro-1H-pyrrolizin-7a(5H)-ylacetic acid hydrochloride

  • MDL number MFCD09032985
  • Cas Number : 124655-63-6
  • Molecular Weight : 205.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA10073
Availability details for
Quantity
  • Availability
  • Add to Cart