ATC311765419 Atlantic Research Chemicals Ltd

4-(Acetylamino)butanoic acid

  • MDL number MFCD00020552
  • Cas Number : 3025-96-5
  • Molecular Weight : 145.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA10117
Availability details for
Quantity
  • Availability
  • Add to Cart