ATC311765450 Atlantic Research Chemicals Ltd

3-Ethoxypropanoic acid

  • MDL number MFCD00058997
  • Cas Number : 4324-38-3
  • Molecular Weight : 118.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA10148
Availability details for
Quantity
  • Availability
  • Add to Cart