ATC311765480 Atlantic Research Chemicals Ltd

3-(3-Fluorophenyl)acryloyl chloride

  • MDL number MFCD03424739
  • Cas Number : 39098-87-8
  • Molecular Weight : 184.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CC00118
Availability details for
Quantity
  • Availability
  • Add to Cart