ATC311765503 Atlantic Research Chemicals Ltd

5-Bromonicotinoyl chloride

  • MDL number MFCD00173853
  • Cas Number : 39620-02-5
  • Molecular Weight : 220.453

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=CC00216
Availability details for
Quantity
  • Availability
  • Add to Cart