ATC311765510 Atlantic Research Chemicals Ltd

1-(4-Chlorophenyl)cyclopropanecarboxylic acid

  • MDL number MFCD00001289
  • Cas Number : 72934-37-3
  • Molecular Weight : 196.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA09937
Availability details for
Quantity
  • Availability
  • Add to Cart