ATC311765540 Atlantic Research Chemicals Ltd

Quinoline-3-carbonyl chloride

  • MDL number MFCD11100097
  • Cas Number : 84741-86-6
  • Molecular Weight : 191.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CC02601
Availability details for
Quantity
  • Availability
  • Add to Cart