ATC311765547 Atlantic Research Chemicals Ltd

4-Benzylmorpholine-2-carbonyl chloride hydrochloride

  • MDL number MFCD03407323
  • Cas Number : 135072-14-9
  • Molecular Weight : 276.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CC04101
Availability details for
Quantity
  • Availability
  • Add to Cart