ATC311765623 Atlantic Research Chemicals Ltd

2,2-Dimethylcyclopropanecarboxylic acid

  • MDL number MFCD04972493
  • Cas Number : 75885-59-5
  • Molecular Weight : 114.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=CA09949
Availability details for
Quantity
  • Availability
  • Add to Cart