ATC311765637 Atlantic Research Chemicals Ltd

Ethyl (4-Aminophenyl)acetate

  • MDL number MFCD00017569
  • Cas Number : 5438-70-0
  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES00115
Availability details for
Quantity
  • Availability
  • Add to Cart