ATC311765648 Atlantic Research Chemicals Ltd

Methyl 3,5-Diisopropoxybenzoate

  • MDL number MFCD08703610
  • Cas Number : 94169-62-7
  • Molecular Weight : 252.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES00124
Availability details for
Quantity
  • Availability
  • Add to Cart