ATC311765659 Atlantic Research Chemicals Ltd

Methyl 4-Amino-3-iodo-5-methylbenzoate

  • MDL number MFCD10703532
  • Cas Number : 180624-11-7
  • Molecular Weight : 291.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES00135
Availability details for
Quantity
  • Availability
  • Add to Cart