ATC311765660 Atlantic Research Chemicals Ltd

Methyl 2-Methyl-4-nitrobenzoate

  • MDL number MFCD03618407
  • Cas Number : 62621-09-4
  • Molecular Weight : 195.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES00136
Availability details for
Quantity
  • Availability
  • Add to Cart