ATC311765662 Atlantic Research Chemicals Ltd

Ethyl (4-Hydrazinophenyl)acetate hydrochloride

  • MDL number MFCD10703534
  • Cas Number : 137402-61-0
  • Molecular Weight : 230.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES00138
Availability details for
Quantity
  • Availability
  • Add to Cart