ATC311765666 Atlantic Research Chemicals Ltd

Methyl 3-Hydroxy-4-methylbenzoate

  • MDL number MFCD00213808
  • Cas Number : 3556-86-3
  • Molecular Weight : 166.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES00142
Availability details for
Quantity
  • Availability
  • Add to Cart