ATC311765671 Atlantic Research Chemicals Ltd

Dimethyl 2-Hydroxyterephthalate

  • MDL number MFCD08445666
  • Cas Number : 6342-72-9
  • Molecular Weight : 210.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES00147
Availability details for
Quantity
  • Availability
  • Add to Cart