ATC311765675 Atlantic Research Chemicals Ltd

2-Hydroxy-4-(methoxycarbonyl)benzoic acid

  • MDL number MFCD11100099
  • Cas Number : 29602-00-4
  • Molecular Weight : 196.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES00151
Availability details for
Quantity
  • Availability
  • Add to Cart