ATC311765686 Atlantic Research Chemicals Ltd

Methyl 3-Methyl-4-nitrobenzoate

  • MDL number MFCD00085640
  • Cas Number : 24078-21-5
  • Molecular Weight : 195.176

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=ES00162
Availability details for
Quantity
  • Availability
  • Add to Cart