ATC311765712 Atlantic Research Chemicals Ltd

Methyl 4-Chloro-2-hydroxybenzoate

  • MDL number MFCD03407436
  • Cas Number : 22717-55-1
  • Molecular Weight : 186.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES00188
Availability details for
Quantity
  • Availability
  • Add to Cart