ATC311765733 Atlantic Research Chemicals Ltd

Methyl 3-Amino-2,6-dichloroisonicotinate

  • MDL number MFCD07437844
  • Cas Number : 883107-62-8
  • Molecular Weight : 221.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES00212
Availability details for
Quantity
  • Availability
  • Add to Cart