ATC311765745 Atlantic Research Chemicals Ltd

Ethyl 2-Chloroisonicotinate

  • MDL number MFCD04114259
  • Cas Number : 54453-93-9
  • Molecular Weight : 185.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES00224
Availability details for
Quantity
  • Availability
  • Add to Cart