ATC311765756 Atlantic Research Chemicals Ltd

Methyl 3-Aminopyridine-2-carboxylate

  • MDL number MFCD08695602
  • Cas Number : 36052-27-4
  • Molecular Weight : 152.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES00235
Availability details for
Quantity
  • Availability
  • Add to Cart