ATC311765864 Atlantic Research Chemicals Ltd

2-tert-Butyl 4-Ethyl 3,5-dimethyl-1H-pyrrole-2,4-dicarboxylate

  • MDL number MFCD02179394
  • Cas Number : 86770-31-2
  • Molecular Weight : 267.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES00511
Availability details for
Quantity
  • Availability
  • Add to Cart