ATC311765910 Atlantic Research Chemicals Ltd

Ethyl 5-Thien-2-yl-1H-pyrazole-3-carboxylate

  • MDL number MFCD07772880
  • Cas Number : 121195-03-7
  • Molecular Weight : 222.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES00906
Availability details for
Quantity
  • Availability
  • Add to Cart