ATC311765916 Atlantic Research Chemicals Ltd

Ethyl 5-Amino-1H-pyrazole-3-carboxylate

  • MDL number MFCD03468385
  • Cas Number : 105434-90-0
  • Molecular Weight : 155.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES00912
Availability details for
Quantity
  • Availability
  • Add to Cart