ATC311765926 Atlantic Research Chemicals Ltd

Ethyl 4-Chloro-2-(trifluoromethyl)pyrimidine-5-carboxylate

  • MDL number MFCD00173897
  • Cas Number : 720-01-4
  • Molecular Weight : 254.594

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=ES01203
Availability details for
Quantity
  • Availability
  • Add to Cart