ATC311765941 Atlantic Research Chemicals Ltd

Methyl 6-Oxo-1,6-dihydropyridazine-3-carboxylate

  • MDL number MFCD08166726
  • Cas Number : 63001-30-9
  • Molecular Weight : 154.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES01302
Availability details for
Quantity
  • Availability
  • Add to Cart