ATC311765944 Atlantic Research Chemicals Ltd

Ethyl 6-Chloropyridazine-3-carboxylate

  • MDL number MFCD09908168
  • Cas Number : 75680-92-1
  • Molecular Weight : 186.597

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=ES01305
Availability details for
Quantity
  • Availability
  • Add to Cart