ATC311766001 Atlantic Research Chemicals Ltd

Ethyl 1-Pyrimidin-2-ylpiperidine-4-carboxylate

  • MDL number MFCD01456221
  • Cas Number : 111247-60-0
  • Molecular Weight : 235.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES03702
Availability details for
Quantity
  • Availability
  • Add to Cart