ATC311766046 Atlantic Research Chemicals Ltd

Methyl 4-Hydroxycyclohexanecarboxylate

  • MDL number MFCD00729087
  • Cas Number : 17449-76-2
  • Molecular Weight : 158.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES09908
Availability details for
Quantity
  • Availability
  • Add to Cart