ATC311766054 Atlantic Research Chemicals Ltd

Ethyl Cyclobutylideneacetate

  • MDL number MFCD11977311
  • Cas Number : 27741-65-7
  • Molecular Weight : 140.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES09916
Availability details for
Quantity
  • Availability
  • Add to Cart