ATC311766122 Atlantic Research Chemicals Ltd

Diethyl 2-Fluoro-2-methylmalonate

  • MDL number MFCD03844748
  • Cas Number : 16519-02-1
  • Molecular Weight : 192.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=ES10104
Availability details for
Quantity
  • Availability
  • Add to Cart