ATC311766172 Atlantic Research Chemicals Ltd

tert-Butyl 4-Aminobutyl(methyl)carbamate

  • MDL number MFCD03001681
  • Cas Number : 144222-23-1
  • Molecular Weight : 202.296

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=ES10154
Availability details for
Quantity
  • Availability
  • Add to Cart