ATC311766271 Atlantic Research Chemicals Ltd

2-(4-Bromophenoxy)-6-(trifluoromethyl)pyridine

  • MDL number MFCD10703540
  • Cas Number : 1086376-15-9
  • Molecular Weight : 318.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=HA00171
Availability details for
Quantity
  • Availability
  • Add to Cart