ATC311766285 Atlantic Research Chemicals Ltd

4-(5-Bromo-2-chlorobenzyl)phenyl ethyl ether

  • MDL number MFCD11042292
  • Cas Number : 461432-23-5
  • Molecular Weight : 325.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=HA00185
Availability details for
Quantity
  • Availability
  • Add to Cart