ATC311766310 Atlantic Research Chemicals Ltd

2,3,4,6-Tetrachloropyridine

  • MDL number MFCD01312169
  • Cas Number : 14121-36-9
  • Molecular Weight : 216.88

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=HA002104
Availability details for
Quantity
  • Availability
  • Add to Cart