ATC311766363 Atlantic Research Chemicals Ltd

2-Chloro-3-nitro-6-(trifluoromethyl)pyridine

  • MDL number MFCD09864674
  • Cas Number : 117519-08-1
  • Molecular Weight : 226.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=HA002155
Availability details for
Quantity
  • Availability
  • Add to Cart