ATC311766393 Atlantic Research Chemicals Ltd

2-Iodo-6-methylpyridine

  • MDL number MFCD11044364
  • Cas Number : 62674-71-9
  • Molecular Weight : 219.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=HA002182
Availability details for
Quantity
  • Availability
  • Add to Cart