ATC311766398 Atlantic Research Chemicals Ltd

2-Fluoro-6-methyl-5-nitropyridine

  • MDL number MFCD03095071
  • Cas Number : 18605-16-8
  • Molecular Weight : 156.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=HA002187
Availability details for
Quantity
  • Availability
  • Add to Cart