ATC311766415 Atlantic Research Chemicals Ltd

2-(Difluoromethoxy)-6-fluoropyridine

  • MDL number MFCD09258966
  • Cas Number : 947534-62-5
  • Molecular Weight : 163.098

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=HA002202
Availability details for
Quantity
  • Availability
  • Add to Cart