ATC311766427 Atlantic Research Chemicals Ltd

2-Iodo-4-methylpyridine

  • MDL number MFCD12963767
  • Cas Number : 22282-60-6
  • Molecular Weight : 219.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=HA002213
Availability details for
Quantity
  • Availability
  • Add to Cart