ATC311766428 Atlantic Research Chemicals Ltd

4-Bromo-1-methylpyridin-2(1H)-one

  • MDL number MFCD11226870
  • Cas Number : 214342-63-9
  • Molecular Weight : 188.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=HA002214
Availability details for
Quantity
  • Availability
  • Add to Cart