ATC311766470 Atlantic Research Chemicals Ltd

6-Chloro-2-iodo-3-(trifluoromethyl)pyridine

  • MDL number MFCD08235251
  • Cas Number : 518057-64-2
  • Molecular Weight : 307.436

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=HA00244
Availability details for
Quantity
  • Availability
  • Add to Cart