ATC311766475 Atlantic Research Chemicals Ltd

5-Chloro-2-hydroxy-3-methylpyridine

  • MDL number MFCD00126916
  • Cas Number : 58498-61-6
  • Molecular Weight : 143.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=HA00250
Availability details for
Quantity
  • Availability
  • Add to Cart