ATC311766506 Atlantic Research Chemicals Ltd

2-Fluoro-5-(trifluoromethyl)pyridine

  • MDL number MFCD07437945
  • Cas Number : 69045-82-5
  • Molecular Weight : 165.089

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin United Kingdom
Product Link https://atlantic-chemicals.com/product/?code=HA00281
Availability details for
Quantity
  • Availability
  • Add to Cart