ATC311766534 Atlantic Research Chemicals Ltd

2-Bromo-5-phenylthiophene

  • MDL number MFCD00159575
  • Cas Number : 29488-24-2
  • Molecular Weight : 239.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://atlantic-chemicals.com/product/?code=HA00310
Availability details for
Quantity
  • Availability
  • Add to Cart